Assay Details
| Gene Symbol: GSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015630.1 | NP_001015630.1 | 4 - 5 | 435 | 98 | |
| GenBank mRNA | BC109713.1 | - | 4 - 5 | 391 | 98 | |
| BT020647.1 | - | 4 - 5 | 435 | 98 | ||
More Information
Set Membership: |
|

GSS
Genomic Map