Assay Details
| Gene Symbol: ACADSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017933.1 | NP_001017933.1 | 5 - 6 | 832 | 62 | |
| GenBank mRNA | BC122698.1 | - | 5 - 6 | 775 | 62 | |
| BT020635.1 | - | 5 - 6 | 832 | 62 | ||
More Information
Set Membership: |
|

ACADSB
Genomic Map