Assay Details
| Gene Symbol: CTSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033617.2 | NP_001028789.1 | 2 - 3 | 383 | 73 | |
| GenBank mRNA | AB060542.1 | - | 2 - 3 | 311 | 73 | |
| BC102115.1 | - | 2 - 3 | 384 | 73 | ||
More Information
Set Membership: |
|

CTSC
Genomic Map