Assay Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034245.2 | NP_001029417.1 | 4 - 5 | 436 | 137 | |
| GenBank mRNA | BC103092.1 | - | 4 - 5 | 437 | 137 | |
More Information
Set Membership: |
|

MAGOH
Genomic Map