Assay Details
| Gene Symbol: CCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034340.2 | NP_001029512.1 | 3 - 4 | 851 | 116 | |
| GenBank mRNA | BC102934.1 | - | 3 - 4 | 852 | 116 | |
More Information
Set Membership: |
|

CCN1
Genomic Map