Assay Details
| Gene Symbol: WBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034346.1 | NP_001029518.1 | 2 - 3 | 316 | 80 | |
| GenBank mRNA | BC103245.1 | - | 2 - 3 | 316 | 80 | |
More Information
Set Membership: |
|

WBP1
Genomic Map