Assay Details
| Gene Symbol: RBM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303463.1 | NP_001290392.1 | 1 - 2 | 88 | 89 | |
| NM_001303480.1 | NP_001290409.1 | 1 - 2 | 88 | 89 | ||
| GenBank mRNA | BC103473.1 | - | 1 - 2 | 81 | 89 | |
| DT860071.1 | - | 1 - 2 | 74 | 89 | ||
| EE239224.1 | - | 1 - 2 | 48 | 89 | ||
| EH156034.1 | - | 1 - 2 | 89 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103473.1, NM_001303463.1, NM_001303480.1 transcripts.
Set Membership: |
|

RBM3
Genomic Map