Assay Details
| Gene Symbol: MFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034397.1 | NP_001029569.1 | 2 - 3 | 411 | 104 | |
| GenBank mRNA | BT020671.1 | - | 2 - 3 | 411 | 104 | |
More Information
Set Membership: |
|

MFAP1
Genomic Map