Assay Details
| Gene Symbol: BPNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034539.2 | NP_001029711.1 | 2 - 3 | 237 | 68 | |
| GenBank mRNA | BC102110.1 | - | 2 - 3 | 239 | 68 | |
| BT030746.2 | - | 2 - 3 | 212 | 68 | ||
More Information
Set Membership: |
|

BPNT1
Genomic Map