Assay Details
| Gene Symbol: MRPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034608.2 | NP_001029780.1 | 1 - 2 | 154 | 67 | |
| GenBank mRNA | BC103149.1 | - | 1 - 2 | 155 | 67 | |
More Information
Set Membership: |
|

MRPL18
Genomic Map