Assay Details
| Gene Symbol: WDR89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034658.2 | NP_001029830.1 | 2 - 3 | 170 | 151 | |
| GenBank mRNA | BC103250.1 | - | 2 - 3 | 171 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103250.1, NM_001034658.2 transcripts.
Set Membership: |
|

WDR89
Genomic Map