Assay Details
| Gene Symbol: TRAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034661.2 | NP_001029833.1 | 6 - 7 | 948 | 68 | |
| GenBank mRNA | BC102522.1 | - | 6 - 7 | 950 | 68 | |
| DQ319074.1 | - | 5 - 6 | 760 | 68 | ||
| DQ407276.1 | - | 5 - 6 | 760 | 68 | ||
| DQ471669.1 | - | 5 - 6 | 767 | 68 | ||
More Information
Set Membership: |
|

TRAF6
Genomic Map