Assay Details
| Gene Symbol: RCAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034679.1 | NP_001029851.1 | 3 - 4 | 499 | 97 | |
| GenBank mRNA | BC103185.1 | - | 3 - 4 | 499 | 97 | |
| BT030515.1 | - | 3 - 4 | 479 | 97 | ||
More Information
Set Membership: |
|

RCAN1
Genomic Map