Assay Details
| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034699.1 | NP_001029871.1 | 5 - 6 | 650 | 60 | |
| GenBank mRNA | BC103292.1 | - | 5 - 6 | 650 | 60 | |
More Information
Set Membership: |
|

PA2G4
Genomic Map