Assay Details
| Gene Symbol: TPPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034774.2 | NP_001029946.1 | 1 - 2 | 128 | 75 | |
| GenBank mRNA | BC102515.1 | - | 1 - 2 | 129 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102515.1, NM_001034774.2 transcripts.
Set Membership: |
|

TPPP3
Genomic Map