Assay Details
| Gene Symbol: TPPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034774.2 | NP_001029946.1 | 2 - 3 | 317 | 89 | |
| GenBank mRNA | BC102515.1 | - | 2 - 3 | 318 | 89 | |
More Information
Set Membership: |
|

TPPP3
Genomic Map