Assay Details
| Gene Symbol: CMTM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035066.1 | NP_001030238.1 | 1 - 2 | 186 | 68 | |
| GenBank mRNA | BC103390.1 | - | 1 - 2 | 186 | 68 | |
More Information
Set Membership: |
|

CMTM6
Genomic Map