Assay Details
| Gene Symbol: PCYT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035089.1 | NP_001030261.1 | 4 - 5 | 441 | 84 | |
| GenBank mRNA | BC103459.1 | - | 4 - 5 | 432 | 84 | |
| BT020694.1 | - | 4 - 5 | 441 | 84 | ||
More Information
Set Membership: |
|

PCYT2
Genomic Map