Assay Details
| Gene Symbol: ZSCAN21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035368.2 | NP_001030445.1 | 3 - 4 | 706 | 77 | |
| GenBank mRNA | BC103036.1 | - | 3 - 4 | 707 | 77 | |
More Information
Set Membership: |
|

ZSCAN21
Genomic Map