Assay Details
| Gene Symbol: GOLGA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035484.1 | NP_001030561.1 | 3 - 4 | 393 | 71 | |
| GenBank mRNA | BC120011.1 | - | 4 - 5 | 442 | 71 | |
| BT020714.1 | - | 3 - 4 | 393 | 71 | ||
More Information
Set Membership: |
|

GOLGA7
Genomic Map