Assay Details
| Gene Symbol: NDUFS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038022.1 | NP_001033111.1 | 3 - 4 | 135 | 58 | |
| GenBank mRNA | BC109716.1 | - | 3 - 4 | 140 | 58 | |
| X65020.1 | - | 135 | 58 | |||
More Information
Set Membership: |
|

NDUFS7
Genomic Map