Assay Details
| Gene Symbol: HNRNPAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040522.1 | NP_001035612.1 | 2 - 3 | 450 | 57 | |
| GenBank mRNA | BC102923.1 | - | 2 - 3 | 450 | 57 | |
More Information
Set Membership: |
|

HNRNPAB
Genomic Map