Assay Details
| Gene Symbol: SERTAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040564.2 | NP_001035654.1 | 1 - 2 | 126 | 152 | |
| GenBank mRNA | BC103440.1 | - | 1 - 2 | 127 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103440.1, NM_001040564.2 transcripts.
Set Membership: |
|

SERTAD1
Genomic Map