Assay Details
| Gene Symbol: OAS1Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040606.1 | NP_001035696.1 | 5 - 6 | 1099 | 107 | |
| GenBank mRNA | AY243505.1 | - | 5 - 6 | 1099 | 107 | |
| BC102471.1 | - | 5 - 6 | 1109 | 107 | ||
More Information
Set Membership: |
|

OAS1Y
Genomic Map