Assay Details
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075428.2 | NP_001068896.1 | 2 - 3 | 353 | 62 | |
| GenBank mRNA | BC103042.1 | - | 2 - 3 | 354 | 62 | |
More Information
Set Membership: |
|

MAL
Genomic Map