Assay Details
| Gene Symbol: PLAC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076987.2 | NP_001070455.1 | 1 - 2 | 80 | 86 | |
| GenBank mRNA | BC102588.1 | - | 1 - 2 | 83 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102588.1, NM_001076987.2 transcripts.
Set Membership: |
|

PLAC8B
Genomic Map