Assay Details
| Gene Symbol: PLAC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076987.2 | NP_001070455.1 | 3 - 4 | 352 | 79 | |
| GenBank mRNA | BC102588.1 | - | 3 - 4 | 355 | 79 | |
More Information
Set Membership: |
|

PLAC8B
Genomic Map