Assay Details
| Gene Symbol: COCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077842.1 | NP_001071310.1 | 11 - 12 | 1579 | 57 | |
| GenBank mRNA | BC123841.1 | - | 11 - 12 | 1582 | 57 | |
| BT020705.1 | - | 11 - 12 | 1579 | 57 | ||
More Information
Set Membership: |
|

COCH
Genomic Map