Assay Details
| Gene Symbol: CLIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080218.2 | NP_001073687.1 | 5 - 6 | 1036 | 85 | |
| GenBank mRNA | BC103261.1 | - | 5 - 6 | 1037 | 85 | |
More Information
Set Membership: |
|

CLIC4
Genomic Map