Assay Details
| Gene Symbol: AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173889.1 | NP_776314.1 | 6 - 7 | 725 | 58 | |
| GenBank mRNA | BC112613.1 | - | 6 - 7 | 718 | 58 | |
| BT025476.1 | - | 6 - 7 | 734 | 58 | ||
| M16225.1 | - | 6 - 7 | 725 | 58 | ||
More Information
Set Membership: |
|

AK2
Genomic Map