Assay Details
| Gene Symbol: CA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173897.1 | NP_776322.1 | 6 - 7 | 614 | 56 | |
| GenBank mRNA | BC142534.1 | - | 6 - 7 | 629 | 56 | |
| U58870.1 | - | 6 - 7 | 614 | 56 | ||
More Information
Set Membership: |
|

CA4
Genomic Map