Assay Details
| Gene Symbol: CPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173903.4 | NP_776328.2 | 5 - 6 | 1213 | 62 | |
| GenBank mRNA | CB437028.1 | - | 1 - 2 | 172 | 62 | |
| EW682509.1 | - | 4 - 5 | 424 | 62 | ||
| MK423995.1 | - | 1 - 2 | 132 | 62 | ||
| X04411.1 | - | 844 | 62 | |||
More Information
Set Membership: |
|

CPE
Genomic Map