Assay Details
| Gene Symbol: CRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173904.2 | NP_776329.1 | 2 - 3 | 185 | 54 | |
| GenBank mRNA | AF154123.1 | - | 2 - 3 | 185 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173904.2 | NP_776329.1 | 2 - 3 | 185 | 54 | |
| GenBank mRNA | AF154123.1 | - | 2 - 3 | 185 | 54 | |
Set Membership: |
|