Assay Details
| Gene Symbol: DDO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173908.2 | NP_776333.1 | 1 - 2 | 55 | 65 | |
| GenBank mRNA | X95310.1 | - | 55 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_173908.2, X95310.1 transcripts.
Set Membership: |
|

DDO
Genomic Map