Assay Details
| Gene Symbol: GAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173914.2 | NP_776339.1 | 3 - 4 | 285 | 71 | |
| GenBank mRNA | BC126798.1 | - | 3 - 4 | 326 | 71 | |
| FJ869997.1 | - | 2 - 3 | 117 | 71 | ||
| MK309336.1 | - | 1 - 2 | 62 | 71 | ||
| X12582.1 | - | 285 | 71 | |||
More Information
Set Membership: |
|

GAL
Genomic Map