Assay Details
| Gene Symbol: IL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173921.2 | NP_776346.1 | 2 - 3 | 250 | 71 | |
| GenBank mRNA | EU276069.1 | - | 2 - 3 | 220 | 71 | |
| M77120.1 | - | 2 - 3 | 250 | 71 | ||
More Information
Set Membership: |
|

IL4
Genomic Map