Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173925.2 | NP_776350.1 | 3 - 4 | 359 | 155 | |
| GenBank mRNA | AF232704.1 | - | 3 - 4 | 359 | 155 | |
| BC103310.1 | - | 3 - 4 | 348 | 154 | ||
| JN559767.1 | - | 3 - 4 | 330 | 154 | ||
More Information
Set Membership: |
|

CXCL8
Genomic Map