Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173931.1 | NP_776356.1 | 1 - 2 | 166 | 63 | |
| GenBank mRNA | U65394.1 | - | 1 - 2 | 166 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_173931.1 transcripts.
Set Membership: |
|

LIF
Genomic Map