Assay Details
| Gene Symbol: MX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173940.2 | NP_776365.1 | 13 - 14 | 1814 | 80 | |
| GenBank mRNA | AB060169.1 | - | 12 - 13 | 2190 | 80 | |
| AF047692.1 | - | 12 - 13 | 1736 | 80 | ||
| BC118109.1 | - | 13 - 14 | 1803 | 80 | ||
| JQ766265.1 | - | 12 - 13 | 1716 | 80 | ||
| U88329.1 | - | 13 - 14 | 1814 | 80 | ||
More Information
Set Membership: |
|

MX1
Genomic Map