Assay Details
| Gene Symbol: CPSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173942.2 | NP_776367.1 | 4 - 5 | 538 | 81 | |
| GenBank mRNA | U96448.1 | - | 4 - 5 | 538 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPSF4| Gene Symbol: CPSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173942.2 | NP_776367.1 | 4 - 5 | 538 | 81 | |
| GenBank mRNA | U96448.1 | - | 4 - 5 | 538 | 81 | |
Set Membership: |
|