Assay Details
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173947.2 | NP_776372.1 | 1 - 2 | 109 | 132 | |
| GenBank mRNA | U67279.1 | - | 1 - 2 | 109 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_173947.2 transcripts.
Set Membership: |
|

OMD
Genomic Map