Assay Details
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173947.2 | NP_776372.1 | 3 - 4 | 1402 | 111 | |
| GenBank mRNA | U67279.1 | - | 3 - 4 | 1402 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173947.2 | NP_776372.1 | 3 - 4 | 1402 | 111 | |
| GenBank mRNA | U67279.1 | - | 3 - 4 | 1402 | 111 | |
Set Membership: |
|