Assay Details
| Gene Symbol: ACO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173977.3 | NP_776402.1 | 12 - 13 | 1509 | 64 | |
| GenBank mRNA | BC102642.1 | - | 12 - 13 | 1510 | 64 | |
| Z49931.1 | - | 1510 | 64 | |||
More Information
Set Membership: |
|

ACO2
Genomic Map