Assay Details
| Gene Symbol: AGER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173982.3 | NP_776407.1 | 4 - 5 | 428 | 103 | |
| GenBank mRNA | BC120127.1 | - | 4 - 5 | 425 | 103 | |
| GU139385.1 | - | 2 - 3 | 794 | 103 | ||
| GU139387.1 | - | 3 - 4 | 614 | 103 | ||
| GU139390.1 | - | 3 - 4 | 615 | 103 | ||
| GU139391.1 | - | 3 - 4 | 615 | 103 | ||
| GU139392.1 | - | 3 - 4 | 615 | 103 | ||
| GU139393.1 | - | 3 - 4 | 616 | 103 | ||
| GU139394.1 | - | 3 - 4 | 614 | 103 | ||
| GU139397.1 | - | 4 - 5 | 103 | |||
| M91212.1 | - | 4 - 5 | 431 | 103 | ||
More Information
Set Membership: |
|

AGER
Genomic Map