Assay Details
| Gene Symbol: CATHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174001.2 | NP_776426.1 | 3 - 4 | 391 | 92 | |
| GenBank mRNA | BC134776.1 | - | 3 - 4 | 392 | 92 | |
| BC142014.1 | - | 3 - 4 | 395 | 92 | ||
| L42977.1 | - | 3 - 4 | 393 | 92 | ||
More Information
Set Membership: |
|

CATHL3
Genomic Map