Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174007.1 | NP_776432.1 | 1 - 2 | 152 | 102 | |
| GenBank mRNA | AF399641.1 | - | 1 - 2 | 81 | 102 | |
| BC142112.1 | - | 1 - 2 | 152 | 102 | ||
| S67954.1 | - | 1 - 2 | 152 | 102 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map