Assay Details
| Gene Symbol: CD247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174012.2 | NP_776437.1 | 3 - 4 | 352 | 54 | |
| GenBank mRNA | BC119996.1 | - | 3 - 4 | 291 | 54 | |
| U25688.1 | - | 3 - 4 | 352 | 54 | ||
More Information
Set Membership: |
|

CD247
Genomic Map