Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174029.1 | NP_776454.1 | 1 - 2 | 310 | 93 | |
| GenBank mRNA | AB098964.1 | - | 1 - 2 | 291 | 93 | |
| AB099041.1 | - | 1 - 2 | 291 | 93 | ||
| BC109629.1 | - | 1 - 2 | 311 | 93 | ||
| Y10811.1 | - | 310 | 93 | |||
More Information
Set Membership: |
|

CST3
Genomic Map