Assay Details
| Gene Symbol: CCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174030.2 | NP_776455.1 | 4 - 5 | 957 | 123 | |
| GenBank mRNA | AB098897.1 | - | 1 - 2 | 82 | 123 | |
| AF000137.1 | - | 4 - 5 | 957 | 123 | ||
| BC113279.1 | - | 4 - 5 | 817 | 123 | ||
More Information
Set Membership: |
|

CCN2
Genomic Map