Assay Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174032.2 | NP_776457.1 | 5 - 6 | 706 | 77 | |
| GenBank mRNA | BC102312.1 | - | 5 - 6 | 752 | 77 | |
| BC151425.1 | - | 5 - 6 | 706 | 77 | ||
| BT021022.1 | - | 5 - 6 | 682 | 77 | ||
| X91755.1 | - | 706 | 77 | |||
More Information
Set Membership: |
|

CTSV
Genomic Map